missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rifampicin-d11 (4-methyl-d3-1-piperazinyl-d8), CDN
Click to view available options
Quantity:
0.005 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Rifampicin-D11 (4-methyl-1-piperazinyl-D11) |
| Molecular Formula | C43 D11 H47 N4 O12 |
| Quantity | 0.005 g |
| Synonym | Rifampicin-d11 (4-methyl-d3-1-piperazinyl-d8), Rifampicin-D11 (4-methyl-1-piperazinyl-D11), Rifampicin-d11, (2 S,12 Z,14 E,16 S,17 S,18 R,19 R,20 R,21 S,22 R,23 S,24 E)-5,6,9,17,19-Pentahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-8-[[[4-(methyl-d3)piperazin-1-yl-2,2,3,3,5,5,6,6-d8]imino]methyl]-1,11-dioxo-1,2-dihydro-2,7-(epoxypentadeca[1,11,13 ]trienimino)naphto[2,1-b]furan-21-yl acetate, 3-[[[4-(Methyl-d3)-1-piperazinyl-2,2,3,3,5,5,6,6-d8]imino]methyl]rifamycin, 5,6,9,17,19,21-Hexahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-8-[N-[4-(methyl-d3)-1-piperazinyl-2,2,3,3,5,5,6,6-d8]formimidoyl]-2,7-(epoxypentadeca[1,11,13 ]trienimino)-naphtho[2,1-b]furan-1,11(2 H)-dione 21-acetate |
| InChI Formula | InChI=1 S/C43H58N4O12/c1-21-12-11-13-22(2)42(55)45-33-28(20-44-47-17-15-46(9)16-18-47)37(52)30-31(38(33)53)36(51)26(6)40-32(30)41(54)43(8,59-40)57-19-14-29(56-10)23(3)39(58-27(7)48)25(5)35(50)24(4)34(21)49/h11-14,19-21,23-25,29,34-35,39,49-53 H,15-18H2,1-10H3,(H,45,55)/b12-11+,19-14+,22-13-,44-20+/t21-,23+,24+,25+,29-,34-,35+,39+,43-/m0/s1/i9D3,15D2,16D2,17D2,18D2 |
| SMILES | [2 H]C([2 H])([2 H])N1C([2 H])([2 H])C([2 H])([2 H])N(\N=C\c2c(O)c3c4C(=O)[C@@]5(C)O\C=C\[C@H](OC)[C@@H](C)[C@@H](OC(=O)C)[C@H](C)[C@H](O)[C@H](C)[C@@H](O)[C@@H](C)\C=C\C=C(\C)/C(=O)Nc2c(O)c3c(O)c(C)c4O5)C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | [(7 S,9 E,11 S,12 R,13 S,14 R,15 R,16 R,17 S,18 S,19 E,21 Z)-2,15,17,27,29-pentahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-26-[(E)-[2,2,3,3,5,5,6,6-octadeuterio-4-(trideuteriomethyl)piperazin-1-yl]iminomethyl]-6,23-dioxo-8,30-dioxa-24-azatetracyclo[23.3.1.14,7.05,28 ]triaconta-1(28),2,4,9,19,21,25(29),26-octaen-13-yl] acetate |
| Molecular Weight (g/mol) | 834.008 |
| Formula Weight | 833.4742 g/mol |
| Percent Purity | 98 atom % D, min 95% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction