missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rift Valley Fever Virus Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41082AF532
This item is not returnable.
View return policy
Description
Rift Valley Fever Virus Polyclonal antibody specifically detects Rift Valley Fever Virus in Virus samples. It is validated for ELISA
Specifications
| Rift Valley Fever Virus | |
| Polyclonal | |
| ELISA | |
| Glycoprotein, RVFV_sM_gp1 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of the RVF virus. The immunogen is located within amino acids 960 - 1010 of Rift Valley Fever Virus. Amino Acid Squence: FERGSLPQTRNDKTFAASK | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 9538296 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction