missing translation for 'onlineSavingsMsg'
Learn More

Rimsulfuron, TRC

Product Code. 30425759
Change view
Click to view available options
Quantity:
100 mg
500 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30425759 5 g
30423821 500 mg
30419318 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425759 Supplier Toronto Research Chemicals Supplier No. TRCR4235005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122931-48-0
Molecular Formula C14 H17 N5 O7 S2
Molecular Weight (g/mol) 431.44
Synonym DPX-E9636, N-[(4,6-dimethoxypyrimidin-2-yl)aminocarbonyl]-3-(ethylsulfonyl)-2-pyridinesulfonamide
IUPAC Name 1-(4,6-dimethoxypyrimidin-2-yl)-3-(3-ethylsulfonylpyridin-2-yl)sulfonylurea
SMILES CCS(=O)(=O)c1cccnc1S(=O)(=O)NC(=O)Nc2nc(OC)cc(OC)n2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C14H17N5O7S2/c1-4-27(21,22)9-6-5-7-15-12(9)28(23,24)19-14(20)18-13-16-10(25-2)8-11(17-13)26-3/h5-8H,4H2,1-3H3,(H2,16,17,18,19,20)
Formula Weight 431.0569
Chemical Name or Material Rimsulfuron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.