missing translation for 'onlineSavingsMsg'
Learn More

Risedronic Acid, TRC

Product Code. 30136052
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136052 2.5 g 2.5g
30137948 250 mg 250mg
30008828 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136052 Supplier Toronto Research Chemicals Supplier No. TRCR5215052.5G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105462-24-6
Molecular Formula C7 H11 N O7 P2
Molecular Weight (g/mol) 283.11
Synonym Phosphonic acid, P,P'-[1-hydroxy-2-(3-pyridinyl)ethylidene]bis-, Phosphonic acid, [1-hydroxy-2-(3-pyridinyl)ethylidene]bis- (9CI), P,P'-[1-Hydroxy-2-(3-pyridinyl)ethylidene]bis[phosphonic acid], 1-Hydroxy-2-(3-pyridinyl)ethylidene bisphosphonic acid, 1-Hydroxy-2-(3-pyridyl)ethylidene-1,1-diphosphonic acid, 1-Hydroxy-2-pyrid-3-ylethylidene-1,1-bisphosphonic acid, BPH 2, NE 58019, Risedronate, Risedronic acid
IUPAC Name (1-hydroxy-1-phosphono-2-pyridin-3-ylethyl)phosphonic acid
SMILES OC(Cc1cccnc1)(P(=O)(O)O)P(=O)(O)O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H11NO7P2/c9-7(16(10,11)12,17(13,14)15)4-6-2-1-3-8-5-6/h1-3,5,9H,4H2,(H2,10,11,12)(H2,13,14,15)
Formula Weight 283.0011
Percent Purity >95%
Chemical Name or Material Risedronic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.