missing translation for 'onlineSavingsMsg'
Learn More

Ritalinic Acid, Mikromol™

Product Code. 30050138
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050138 100 mg 100mg
30052468 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050138 Supplier Mikromol Supplier No. MM1031.04
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ritalinic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ritalinic Acid
CAS 19395-41-6
Analyte or Component Names Ritalinic Acid
Shipping Condition Room Temperature
Molecular Formula C13 H17 N O2
Certifications/Compliance ISO 17025
Synonym Ritalinic Acid, Methylphenidate Related Compound A
InChI Formula InChI=1S/C13H17NO2/c15-13(16)12(10-6-2-1-3-7-10)11-8-4-5-9-14-11/h1-3,6-7,11-12,14H,4-5,8-9H2,(H,15,16)
SMILES OC(=O)C(C1CCCCN1)c2ccccc2
IUPAC Name 2-phenyl-2-piperidin-2-ylacetic acid
Molecular Weight (g/mol) 219.28
Quantity 100 mg
Formula Weight 219.1259 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.