missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rizatriptan benzoate

Product Code. 16544224 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16544224 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16544224 Supplier Thermo Scientific Chemicals Supplier No. 462940010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 145202-66-0
Melting Point 178.0°C to 180.0°C
Molecular Formula C22H25N5O2
Quantity 1 g
Solubility Information Solubility aq. soln.: 53 g/L Other solubilities: soluble in methanol, slightly soluble in ethanol, insoluble in ether
InChI Key JPRXYLQNJJVCMZ-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=CC=C1.CN(C)CCC1=CNC2=CC=C(CN3C=NC=N3)C=C12
IUPAC Name benzoic acid; dimethyl(2-{5-[(1H-1,2,4-triazol-1-yl)methyl]-1H-indol-3-yl}ethyl)amine
Molecular Weight (g/mol) 391.48
Formula Weight 391.47
Chemical Name or Material Rizatriptan benzoate
Show More Show Less
compliance-icons
Product Identifier
  • Rizatriptan benzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Rizatriptan benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.