missing translation for 'onlineSavingsMsg'
Learn More

RN 1747, TRC

Product Code. 30409871
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30409871 250 mg
30413380 50 mg
30409248 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30409871 Supplier Toronto Research Chemicals Supplier No. TRCR637008250MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1024448-59-6
Molecular Formula C17H18ClN3O4S
Molecular Weight (g/mol) 395.87
Synonym RN 1747, 1-(4-Chloro-2-nitrophenyl)sulfonyl-4-benzylpiperazine
IUPAC Name 1-benzyl-4-(4-chloro-2-nitrophenyl)sulfonylpiperazine
SMILES [O-][N+](=O)c1cc(Cl)ccc1S(=O)(=O)N2CCN(Cc3ccccc3)CC2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H18ClN3O4S/c18-15-6-7-17(16(12-15)21(22)23)26(24,25)20-10-8-19(9-11-20)13-14-4-2-1-3-5-14/h1-7,12H,8-11,13H2
Formula Weight 395.0707
Chemical Name or Material 1-(4-Chloro-2-nitrophenyl)sulfonyl-4-benzylpiperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.