missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASE7 Antibody (CL0223) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52883B
This item is not returnable.
View return policy
Description
RNASE7 Monoclonal antibody specifically detects RNASE7 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| RNASE7 | |
| Monoclonal | |
| Biotin | |
| EC 3.1.27, EC 3.1.27.-, EC 3.1.27.5, MGC133220, ribonuclease 7, ribonuclease, RNase A family, 7, RNase 7, SAP-2, Skin-derived antimicrobial protein 2 | |
| This antibody was developed using a recombinant protein derived from Q9H1E1, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 84659 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0223 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction