missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNaseH2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524708
This item is not returnable.
View return policy
Description
RNaseH2B Polyclonal Antibody for Western Blot, IHC (P)
RNASEH2B is composed of a single catalytic subunit (A) and two non-catalytic subunits (B and C) and specifically degrades the RNA of RNA:DNA hybrids. RNASEH2B is the non-catalytic B subunit of RNase H2, which is thought to play a role in DNA replication.
Specifications
| RNaseH2B | |
| Polyclonal | |
| Unconjugated | |
| RNASEH2B | |
| 1110019N06Rik; 2610207P08Rik; AGS2; Aicardi-Goutieres syndrome 2 protein; Deleted in lymphocytic leukemia 8; deleted in lymphocytic leukemia 8 homolog; DLEU8; ribonuclease H2; ribonuclease H2 subunit B; ribonuclease H2, subunit B; ribonuclease HI subunit B; RNase H2 subunit B; Rnaseh2b | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 79621 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q5TBB1 | |
| RNASEH2B | |
| KLH conjugated synthetic peptide between 130-157 amino acids from the central region of human RNASEH2B. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction