missing translation for 'onlineSavingsMsg'
Learn More

Löser™ Roebling™ Inking Ribbon

Product Code. 11900098
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11900098 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11900098 Supplier Löser™ Supplier No. DPN 190
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Compatible with Roebling™ printer DR01/DR02. Robeling™ Inking Ribbon is for use with Osmometer type 13DR.

For use with Roebling™ printer DR01/DR02.and Osmometer type 13DR.

Specifications

Color Black
For Use With (Equipment) Roebling Printer DR01/DR02 with Osmometer Type 13DR
Material Plastic
Weight (Metric) 8 g
For Use With (Application) Printing results with Osmometer type 13DR.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.