Learn More
Rogo Sampaic™ Glass Coverslips
Product Code. 11767394
Change view
Click to view available options
Length (Metric):
18 mm
20 mm
22 mm
24 mm
Width (Metric):
18 mm
20 mm
22 mm
50 mm
60 mm
Quantity:
100/Pk.
1000/Pk.
200/Pk.
Unit Size:
Pack of 100
Pack of 1000
Pack of 200
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Quantity Unit Size
11767394 24 mm 60 mm 100/Pk. Pack of 100
11993564 18 mm 18 mm 1000/Pk. Pack of 1000
11707394 20 mm 20 mm 200/Pk. Pack of 200
11757394 24 mm 50 mm 100/Pk. Pack of 100
11717394 22 mm 22 mm 200/Pk. Pack of 200
11983564 24 mm 60 mm 100/Pk. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11767394 Supplier Rogo Sampaic™ Supplier No. BPD025
  • ONLINE EXCLUSIVE PRICE
    19.98€
    7.19€ / Pack of 100
    Save 12.79€
    64% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Preserve and protect specimens on slides. Rogo Sampaic™ Glass Coverslips are manufactured from borosilicate glass for excellent transparency and prevention of color distortion.
  • Available in a range of sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Product Type Cover Slide
Shape Rectangle
Material Borosilicate Glass
Length (Metric) 24 mm
Width (Metric) 60 mm
Dimensions (L x W) 24 x 60 mm
For Use With (Application) Slide microscopy
Certifications ISO 8255
Quantity 100/Pk.