Learn More
Rogo Sampaic™ Glass Coverslips
Artikelnummer. 11993564
Skift visning
Klik for at se tilgængelige muligheder
Length (Metric):
18 mm
20 mm
22 mm
24 mm
Width (Metric):
18 mm
20 mm
22 mm
50 mm
60 mm
Quantity:
100/Pk.
1000/Pk.
200/Pk.
Pakningsstørrelse:
Pack of 100
Pack of 1000
Pack of 200
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Length (Metric) Width (Metric) Quantity Pakningsstørrelse
11993564 18 mm 18 mm 1000/Pk. Pack of 1000
11707394 20 mm 20 mm 200/Pk. Pack of 200
11757394 24 mm 50 mm 100/Pk. Pack of 100
11717394 22 mm 22 mm 200/Pk. Pack of 200
11767394 24 mm 60 mm 100/Pk. Pack of 100
11983564 24 mm 60 mm 100/Pk. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Artikelnummer. 11993564 Leverandør Rogo Sampaic™ Leverandørnr. BPD016
  • ONLINE EXCLUSIVE PRICE
    21.65€
    16.55€ / Pack of 1000
    Save 5.10€
    23% Off
In Stock
Læg i indkøbskurv
Edge
Læg i indkøbskurv
missing translation for 'validMainframeMsg'
Preserve and protect specimens on slides. Rogo Sampaic™ Glass Coverslips are manufactured from borosilicate glass for excellent transparency and prevention of color distortion.
  • Available in a range of sizes
  • Corrosion-free and resistant to thermal shock
Tekniske data
Product Type Cover Slide
Shape Square
Material Borosilicate Glass
Length (Metric) 18 mm
Width (Metric) 18 mm
Dimensions (L x W) 18 x 18 mm
For Use With (Application) Slide microscopy
Certifications ISO 8255
Quantity 1000/Pk.
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.