missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Roledumab Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

477.00€ - 1204.00€

Specifications

Antigen Roledumab
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30283211
GREENER_CHOICE
View Documents
Invitrogen™
MA559761
100 μg
477.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30282574
View Documents
Invitrogen™
MA559762
1 mg
1204.00€
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Roledumab is a biosimilar that targets RHD. The Rhesus (Rh) blood group system represents one of the most complex and important systems in humans. Two highly homologous genes RHD and RHCE (collectively referred to as RH30 or RHCED) encode the antigens of the Rh blood group system (1-4). These tightly linked genes map to human chromosomal position 1p34.1-1p36. The RHD gene, which is commonly deleted from a large segment of the population, encodes the most potent blood group immunogen, the D antigen (2,5). Rh incompatibility between maternal and fetal blood types results in hemolytic disease of the newborn (HDN), which often results in fetal death (1,2,6). The RHCE gene exists in four allelic forms, and each allele determines the expression of two antigens in Ce, ce, cE, or CE combinations (2,4,5). The RHCED antigens exist as integral membrane proteins with contain 12-transmembrane helices, and maintain erythrocyte membrane integrity. The presentation of the Rh antigenic activity requires the formation of a complex between the RHCED antigens and RHAG (RH50).
Specifications

Specifications

Roledumab
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
LFB-R593
Antibody
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.