Learn More
Invitrogen™ Romilkimab Chimeric Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542185
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt. Isotype of this product is: [VH-VH'-H-Gamma1-VL-VL'-C, kappa] dimer.
Romilkimab is a biosimilar that targets IL13.
Specifications
| Romilkimab Chimeric | |
| Recombinant Monoclonal | |
| Unconjugated | |
| huTBTI3_2_1; SAR156597 | |
| Human IL13/IL4. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, SDS-Page | |
| 1.9 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| VH-VH |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.