missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ ROPN1B 293T Cell Transient Overexpression Lysate

Product Code. 18568301 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18568301 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18568301 Supplier R&D Systems™ Supplier No. H00152015T01
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Product Line Novus
Target ROPN1B
Synonym AKAP-binding sperm protein ropporin, rhophilin associated tail protein 1 B, Rhophilin-associated protein 1 B, ropporin, rhophilin associated protein 1 B, ropporin-1 B
Gene Symbol ROPN1B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.