missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Roquin Antibody [FITC], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB100-655F
This item is not returnable.
View return policy
Description
Roquin Polyclonal antibody specifically detects Roquin in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Roquin | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| FITC | |
| KIAA2025RP5-1198E17.5, RING finger and C3H zinc finger protein 1, ring finger and CCCH-type domains 1, RING finger protein 198, RNF198ring finger and CCCH-type zinc finger domains 1, roquin | |
| The immunogen recognized by this antibody maps to a region between residues 500 and 550 of human Ring Finger and CCCH-type finder domains 1 using the numbering given in entry NP_742068.1 (GeneID 149041). | |
| 0.1 mL | |
| Immunology | |
| 149041 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction