Learn More
BUCHI Rotavapor I-300 Vacuum Controller and Interface
Product Code. 17274694 Shop All Buchi Products
Change view
Click to view available options
Model:
Pro
Standard
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
17274694 Standard Each
17284694 Pro Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17274694 Supplier BUCHI Supplier No. 11I300P40
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
The BUCHI Interface I-300 controls all distillation parameters of a Rotavapor R-300 system including the optional V-300 Vacuum Pump and F-3XX series chillers.
  • Easy plug and play integration into a Rotavapor R-300 system at any time
  • Central control of all process parameters such as rotation speed, temperature (heating, cooling, vapor) and pressure
  • Remote monitoring on smart phones including push notifications and live view
  • Modes: manual, timer, continuous
  • Flexibility of the interface allows easy installation and convenient operation from multiple positions:
    • Rotavapor handle
    • Vacuum pump
    • Fume hood or lab bench wall
  • Stand-alone operation to locally control a central vacuum system
  • Synchronized processing thanks to centrally controlled rotary evaporator, heating bath, speed-controlled vacuum pump and recirculating chiller
  • Unattended operation (optional):
    • Automatic distillation with CloudDest sensor
    • Foam sensor for foaming samples
TRUSTED_SUSTAINABILITY
Specifications
Type Vacuum Tubing
Includes Kit for R-21x/V-7x0
Model Standard