Learn More
Thermo Scientific™ Rotor, 4-Place Swinging Bucket, for Thermo Scientific Sorvall™ Legend™ Mach 1.6™ Centrifuge
Product Code. 10502173 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
4 x 400 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
10502173 4 x 400 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10502173 Supplier Thermo Scientific™ Supplier No. 75002000
  • ONLINE EXCLUSIVE PRICE
    2585.00€
    2470.00€ / Pack of 1
    Save 115.00€
    4% Off
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
For use with Legend Mach 1.6 and 1.6R Centrifuges
  • Rotor body only; buckets and biocontainment lids sold separately
  • Choose rectangular or round buckets
  • Adapters sold separately
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor
Capacity 4 x 400 mL
Max. RCF 4,618 x g
Max. Speed 4,700 rpm
For Use With (Equipment) Legend Mach 1.6 Series