Learn More
Rotronic™ Disposable Sample Cups
Product Code. 11950685
Change view
Click to view available options
Depth (English):
0.55 in.
1.57 in.
Depth (Metric):
1.4 cm
4 cm
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (English) Depth (Metric) unitSize
11950685 0.55 in. 1.4 cm Pack of 100
11940685 1.57 in. 4 cm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11950685 Supplier Rotronic™ Supplier No. PS14
  • ONLINE EXCLUSIVE PRICE
    245.30€
    192.53€ / Pack of 100
    Save 52.77€
    21% Off
In Stock
Add to basket
Edge
Add to basket
Ensures optimum sample volume is introduced into the sample holders.
  • Prevent sample holders from coming into direct contact with product being tested, thereby preventing soiling or cross contamination
  • Provides convenient means of collecting and storing samples
Specifications
Depth (Metric) 1.4 cm
Depth (English) 0.55 in.
Product Type Sample Cup
Disposable Single-use
For Use With (Application) Collecting and Storing samples
For Use With (Equipment) WP-14-S sample holders