Learn More
IKA™ Round Bottom Flask Carrier
Product Code. 15254037 Shop All IKA Products
Change view
Click to view available options
Capacity (Metric):
100 mL
1000 mL
2000 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15254037 1000 mL Each
15188449 100 mL Each
15375051 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15254037 Supplier IKA™ Supplier No. 0025000833
Only null left
Add to basket
Add to basket
Compatible with several IKA stirrers.
  • Round/hollow cylindrical bottom
  • Aluminum anodized
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1000 mL
Includes Without handle
Shape Round Bottom
Temperature (Metric) 180°C
Type Flask Carrier