Learn More
IKA™ Round Bottom Flask Inlay
Product Code. 15224007 Shop All IKA Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
500 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15224007 25 mL Each
15178449 50 mL Each
15213305 10 mL Each
15214037 100 mL Each
15284037 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15224007 Supplier IKA™ Supplier No. 0025000637
Only null left
Add to basket
Add to basket
Round bottom flask inlay constructed of anodized aluminum
  • Round bottom
  • Anodized aluminum construction
  • Temperature resistant
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 25 mL
Diameter (English) 1.7 in.
Diameter (Metric) 44 mm
Shape Round Bottom
Temperature (Metric) 180°C
Type Flask Inlay