missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RP11-195F19.5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00€ - 593.00€
Specifications
| Antigen | RP11-195F19.5 |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18601839
|
Novus Biologicals
NBP2-49609-25ul |
25 μL |
369.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649458
|
Novus Biologicals
NBP2-49609 |
0.1 mL |
593.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RP11-195F19.5 Polyclonal antibody specifically detects RP11-195F19.5 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| RP11-195F19.5 | |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CDNA FLJ50015, HCG2040265, Isoform CRA_a, LOC730098, Novel Protein, Uncharacterized protein, Uncharacterized Protein 13 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PLQETFGAQDMSGMRRYEVALEAEEEIYWGCFYFFPWLRMWRRERSSAHPGEQKLEPLRGLMSCLSSGLGPTPQRSGRGFPRRSPTAAAQPASALK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title