Learn More
RPA1 Mouse anti-Human, Unconjugated, Clone: 1G3F11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67973-1-IG-150UL
 View more versions of this product
Product Code. 17591486
  • 410.00€ / 150µL
Description
The RPA1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human RPA1. This antibody recognizes human, rat antigen. The RPA1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
RPA1
Monoclonal
Unconjugated
150 μL
6117
IgG1
Western Blot
1G3F11
Mouse
Primary
Human, Rat