missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL26L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549955
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total RPL26L1 protein.
This gene encodes a protein that shares high sequence similarity with ribosomal protein L26. It is not currently known whether the encoded protein is a functional ribosomal protein or whether it has evolved a function that is independent of the ribosome. Transcript variants utilizing alternative polyA signals exist. [provided by RefSeq, Jul 2008].
Specifications
| RPL26L1 | |
| Polyclonal | |
| Unconjugated | |
| RPL26L1 | |
| 60S ribosomal protein L26-like 1; Large ribosomal subunit protein uL24-like 1; ribosomal protein L26 homolog; ribosomal protein L26 like 1; ribosomal protein L26 pseudogene 1; RPL26L1; RPL26P1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 307636, 51121, 668405 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UNX3 | |
| Rpl26-ps3, RPL26L1 | |
| Synthesized peptide derived from C-terminal of human RPL26L. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction