missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rpn2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 550.00€
Specifications
| Antigen | Rpn2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230562
|
Novus Biologicals
NBP3-35667-20ul |
20 μL |
190.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30226687
|
Novus Biologicals
NBP3-35667-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rpn2 Polyclonal antibody specifically detects Rpn2 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| Rpn2 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Neuroscience, Ubiquitin Proteasome Pathway | |
| PBS (pH 7.3), 50% glycerol | |
| 5707 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 26S proteasome non-ATPase regulatory subunit 1, 26S proteasome regulatory subunit RPN2, 26S proteasome regulatory subunit S1, 26S proteasome subunit p112, MGC133040, MGC133041, P112, proteasome (prosome, macropain) 26S subunit, non-ATPase, 1, Rpn2, S1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 794-953 of human Rpn2 (NP_002798.2).,, Sequence:, DLKMPKVQYKSNCKPSTFAYPAPLEVPKEKEKEKVSTAVLSITAKAKKKEKEKEKKEEEKMEVDEAEKKEEKEKKKEPEPNFQLLDNPARVMPAQLKVLTMPETCRYQPFKPLSIGGIIILKDTSEDIEELVEPVAAHGPKIEEEEQEPEPPEPFEYIDD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title