Learn More
RPRD1B Antibody (01), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-06202CL1
Product Code. 30153706
  • 801.00€ / 0.10mL
Description
RPRD1B Monoclonal antibody specifically detects RPRD1B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RPRD1B
Monoclonal
CoraFluor 1
Cell cycle-related and expression-elevated protein in tumor, cell-cycle related and expression-elevated protein in tumor, chromosome 20 open reading frame 77, CREPTC20orf77, dJ1057B20.2, DKFZp434P0735, FLJ44520, NET60, regulation of nuclear pre-mRNA domain containing 1B, regulation of nuclear pre-mRNA domain-containing protein 1B
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human RPRD1B (Accession#: AAH33629.1; Met1-Asp326).
0.1 mL
Primary
Human
Purified
Western Blot
01
PBS
Mouse
Protein A purified
RUO
58490
Store at 4°C in the dark. Do not freeze.
IgG2b