Learn More
RPS3 Mouse anti-Human, CoraLite 594, Clone: 2G7H4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66046-100UL
Description
The RPS3 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human RPS3. This antibody recognizes human, mouse, rat antigen. The RPS3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| RPS3 | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 6188 | |
| IgG2a |
| Immunofluorescence | |
| 2G7H4 | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |