Learn More
Invitrogen™ Human RPS6KA2 (RSK3), GST Tag Recombinant Protein
Human RPS6KA2 (RSK3), GST Tag Recombinant Protein
Brand: Invitrogen™ A31518
Additional Details : Weight : 0.01000kg
Description
The purified recombinant full-length protein can be used in a number of different applications, but it has been specifically validated for use with both the LanthaScreen™ & Z'-LYTE™ Assay technologies to identify novel kinase inhibitors. RPS6KA2 is also available in our SelectScreen™ Kinase Profiling Service.
- Validated for use with the LanthaScreen & Z'-LYTE assay technology
- Available in multiple pack sizes for convenient use
- Also available in our SelectSceen Kinase Profiling Service
Specifications
NP_066958.2 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. | |
100μg | |
RUO - research use only | |
p90-RSK3/HU-2/RSK/MAPKAPK1C/S6K-alpha | |
Full Length | |
Recombinant | |
Insect |
Kinase Assay | |
109.9kDa | |
Store at -80°C. Upon first use, aliquot and store at -80°C to avoid multiple freeze/thaws. | |
Full Length, Recombinant, Active | |
RSK (RPS6K) Kinases | |
Human | |
GST Tag |