missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPS6KA4 (MSK2) Recombinant Human Protein

RPS6KA4 (MSK2) is a serine/threonine protein kinase that is essential for the regulation of various cellular processes.

Brand:  Invitrogen™ PV4052

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 10261403

  • 7890.00€ / 1mg
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The RPS6KA4 (MSK2) kinase contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including CREB1 and c-fos.

Specifications

Specifications

NP_003933
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
RPS6KA4 (MSK2)
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
113.1kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
RSK (RPS6K) Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions