Learn More
RPS6KC1 Monoclonal Antibody (OTI10F11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808825S
Description
RPS6KC1 Monoclonal antibody specifically detects RPS6KC1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| RPS6KC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14893 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8487 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI10F11 | |
| Unconjugated | |
| GEMIN2 | |
| 52 kDa ribosomal protein S6 kinase; AA682037; B130003F20Rik; C80612; humS6PKh1; ribosomal protein S6 kinase C1; ribosomal protein S6 kinase delta-1; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 52kDa, polypeptide 1; ribosomal S6 kinase-like protein with two PSK domains 118 kDa protein; RPK118; Rps6kc1; RSKL1; S6K-delta-1; S6PKh1; SPHK1-binding protein | |
| Full length human recombinant protein of human SIP1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.