missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRAS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
217.00€ - 349.00€
Specifications
| Antigen | RRAS |
|---|---|
| Dilution | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18038375
|
Novus Biologicals
NB100-56161SS |
0.025 mL |
217.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18774983
|
Novus Biologicals
NB100-56161 |
0.05 mL |
349.00€
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RRAS Polyclonal specifically detects RRAS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| RRAS | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| Whole antisera with 0.05% Sodium Azide | |
| 6237 | |
| A synthetic peptide made to an internal portion of the mouse RRAS protein (between amino acids 10-75) [UniProt P10833] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat | |
| Oncogene RRAS, p23, ras-related protein R-Ras, related RAS viral (r-ras) oncogene homolog | |
| RRAS | |
| IgG | |
| Unpurified | |
| R-Ras |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title