Learn More
RRM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19686-1-AP-20UL
 View more versions of this product
Product Code. 16899154
  • 145.00€ / 20µL
Description
The antibody is specific to RRM1.
This gene encodes one of two non-identical subunits that constitute ribonucleoside-diphosphate reductase, an enzyme essential for the production of deoxyribonucleotides prior to DNA synthesis in S phase of dividing cells. It is one of several genes located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocrotical carcinoma, and lung, ovarian, and breast cancer. This gene may play a role in maligcies and disease that involve this region.
Specifications
Specifications
RRM1
Polyclonal
Unconjugated
RRM1
R1, ribonucleotide reductase M1, RIR1, RR1, RRM1
Rabbit
Antigen Affinity Chromatography
RUO
20133, 6240, 685579
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunofluorescence
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07742, P23921
RRM1
Peptide
20 μL
Primary
Human, Rat, Mouse