missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RRP15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555134
This item is not returnable.
View return policy
Description
Immunogen sequence: VSEEENLKKT PKKKMKMVTG AVASVLEDEA TDTSDSEGSC GSEKDHFYSD DDAIEADSEG DAEPCDKENE NDGESSVGTN Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 54%.
This gene encodes a protein that co-purifies with human nucleoli. A similar protein in budding yeast is a component of pre-60S ribosomal particles, and is required for the early maturation steps of the 60S subunit.
Specifications
| RRP15 | |
| Polyclonal | |
| Unconjugated | |
| RRP15 | |
| 2810430M08Rik; 5430437H21Rik; CGI-115; KIAA0507; LOC505387 protein; RGD1306106; ribosomal RNA processing 15 homolog; ribosomal RNA processing 15 homolog (S. cerevisiae); ribosomal RNA-processing protein 15; RRP15; RRP15-like protein; RRP15-like protein-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51018 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y3B9 | |
| RRP15 | |
| Recombinant protein corresponding to Human RRP15. Recombinant protein control fragment (Product #RP-94006). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction