missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15329-1-AP-20UL
This item is not returnable.
View return policy
Description
Involved in ribosome biogenesis. .Specifications
| RRS1 | |
| Polyclonal | |
| Unconjugated | |
| RRS1 | |
| KIAA0112, RRR, RRS1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23212, 297784, 59014 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A1A5P2, Q15050, Q9CYH6 | |
| Rrs1 | |
| RRS1 Fusion Protein Ag7030 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction