missing translation for 'onlineSavingsMsg'
Learn More

(RS)-Phenyl(pyridin-2-yl)acetonitrile (Pyronitrile), Mikromol™

Product Code. 30458202
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458202 4 x 25 mg 100mg
30051494 25 mg 25mg
30049276 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30458202 Supplier Mikromol Supplier No. MM0108.0100254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(RS)-Phenyl(pyridin-2-yl)acetonitrile (Pyronitrile), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material a-Phenyl-a-(2-pyridyl)acetonitrile
CAS 5005-36-7
Analyte or Component Names a-Phenyl-a-(2-pyridyl)acetonitrile
Shipping Condition Room Temperature
Molecular Formula C13 H10 N2
Certifications/Compliance ISO 9001
Synonym 2-Pyridineacetonitrile, alpha-phenyl-, 2-Phenyl-2-(2-pyridyl)acetonitrile, 2-Pyridylphenylacetonitrile, NSC 16276, Phenyl(pyridin-2-yl)acetonitrile, alpha-Phenyl-2-pyridineacetonitrile, alpha-Phenyl-alpha-(2-pyridyl)acetonitrile, Disopyramide Impurity D
InChI Formula InChI=1S/C13H10N2/c14-10-12(11-6-2-1-3-7-11)13-8-4-5-9-15-13/h1-9,12H
Solution Type Neat
SMILES N#CC(c1ccccc1)c2ccccn2
IUPAC Name 2-phenyl-2-pyridin-2-ylacetonitrile
Molecular Weight (g/mol) 194.23
Quantity 4 x 25 mg
Formula Weight 194.0844 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.