Learn More
STEMCELL Technologies RSeT™ Feeder-Free Medium
Product Code. 16467933
Encompass
Change view
Click to view available options
Quantity:
500 mL Kit
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467933 500 mL Kit 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16467933 Supplier STEMCELL Technologies Supplier No. 05975
Serum-free medium for naïve-like human pluripotent stem cells
  • Revert primed human embryonic (ES) and induced pluripotent stem (iPS) cells to a naïve-like state and maintain cells in a naïve-like state under hypoxic conditions without bFGF or feeder cells
  • Developed under license from the Weizmann Institute of Science, this serum-free cell culture medium produces robust cultures with features of a naïve-like state, such as tightly packed, domed colonies with refractive edges
  • Human ES/iPS cells cultured in RSeT Feeder-Free Medium show increased expression of key transcripts associated with the naïve-like state, such as KLF17, KLF2, KLF4, and TFCP2L1, and can either be differentiated or converted back to a primed state by culture in mTeSR1 and then differentiated
  • Compatible products that can be used for differentiation include STEMdiff Definitive Endoderm Kit (Catalog #05110), STEMdiff SMADi Neural Induction Kit (Catalog #08581), and STEMdiff Mesoderm Induction Medium (Catalog #05220)
Specifications
Cell Type Pluripotent Stem Cells
For Use With (Application) Cell Culture, Expansion, Maintenance
Quantity 500 mL Kit