Learn More
RSPO3 Monoclonal Antibody (OTI3A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA804635S
Encompass_Preferred
 View more versions of this product
Product Code. 17514283
  • 242.00€ / 30µL
Description
RSPO3 Monoclonal antibody specifically detects RSPO3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
RSPO3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BXY4
Mouse
Affinity chromatography
RUO
84870
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI3A9
Unconjugated
RSPO3
2810459H04Rik; AW742308; Cabriolet; cristin 1; Cristin1; cristin-1; cysteine-rich and single thrombospondin domain-containing protein 1; hPWTSR; hRspo3; Nucleopondin; protein with TSP type-1 repeat; PWTSR; RGD1563246; Roof plate-specific spondin-3; Rspo3; R-spondin 3; R-spondin 3 homolog; R-spondin 3 homolog (Xenopus laevis); R-spondin 3-like protein; R-spondin-3; Thrombospondin type-1 domain-containing protein 2; thrombospondin, type I, domain 2; thrombospondin, type I, domain containing 2; Thsd2; Unknown (protein for MGC:137973)
Human recombinant protein fragment corresponding to amino acids 22-272 of human RSPO3 produced in E.coli.
30 μL