Learn More
RSPO3 Monoclonal Antibody (OTI3A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804635
Description
RSPO3 is a member of the R-spondin family and plays a role in the regulation of Wnt (wingless-type MMTV integration site family)/beta-catenin and Wnt/planar cell polarity (PCP) signaling pathways, which are involved in development, cell growth and disease pathogenesis. RSPO3 has been shown to be involved in regulating the balance between angioblast and blood cell specification during embryonic vasculogenesis and angiogenesis and may be involved in tumor development.Specifications
| RSPO3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BXY4 | |
| RSPO3 | |
| Human recombit protein fragment corresponding to amino acids 22-272 of human RSPO3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3A9 | |
| Unconjugated | |
| RSPO3 | |
| CRISTIN1, PWTSR, THSD2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84870 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.