Learn More
Abeba™ Rubber 9250 Shoes
Product Code. 15445713
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15445713 36 1 pair
15455713 37 1 pair
15465713 38 1 pair
15475713 39 1 pair
15485713 40 1 pair
15495713 41 1 pair
15415723 42 1 pair
15425723 43 1 pair
15435723 44 1 pair
15673208 45 1 pair
15683208 46 1 pair
15693208 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15445713 Supplier Abeba™ Supplier No. 9250/36
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
Acid and base resistant. Rubber sole provides high slip resistance.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Dark blue
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC/li
Color Blue
Material Leather
Size 36
Sole Features Slip-resistant
Sole Material Rubber
Style 9250