Learn More
Bellingham Stanley™ Rubber Gland Seals for Metal End-Cap Polarimeter Tubes
Product Code. 16267350
Change view
Click to view available options
For Use With (Equipment):
Metal End-Cap Polarimeter Tube
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16267350 Metal End-Cap Polarimeter Tube Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16267350 Supplier Bellingham Stanley™ Supplier No. 3521
 Restricted Item
Estimated Shipment: 24-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Rubber Gland Washers for Metal End Cap Tubes & Fitting Tool are spare parts for Bellingham Stanley™ polarimeter tubes.
  • Rubber seal washers for metal-ended glass polarimeter tubes
  • Accessory for Bellingham Stanley polarimeters
  • Supplied with installation tool
  • Pack of 12
TRUSTED_SUSTAINABILITY
Specifications
Type Gland Seal Washer
Material Rubber
For Use With (Equipment) Metal End-Cap Polarimeter Tube
Length (Metric) 15.5 mm