missing translation for 'onlineSavingsMsg'
Learn More

Rubidium Standard: Rb @ 10000 μg/mL in 5% HNO3

Product Code. 30313550
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313550 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 30313550 Supplier VHG Labs Supplier No. VHGTRBN100

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Rubidium
CAS 7440-17-7
Analyte or Component Names Rubidium
Shipping Condition Room Temperature
Molecular Formula Rb
Concentration 10000 μg/mL
Certifications/Compliance ISO 17034
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Synonym Rb
InChI Formula InChI=1 S/Rb
Solution Type Single Solution
SMILES [Rb]
Element Rb
IUPAC Name rubidium
Solvent or Matrix Dilute HNO3
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 85.468
Quantity 100 mL
Formula Weight 84.912
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.