Description
- Good heat/cold insulation on the palm
- Abrasion and cut resistance
- Ergonomic fit
- High flexibility
- Excellent mechanical characteristics
- Good resistance to many chemicals, acids, alkalis, mineral oils and solvents
- High water vapor absorption due to the cotton lining
- Fully coated
- Sandwich lining on palm, cuff
Specifications
Specifications
| Certifications/Compliance | EN: 388 (2 1 2 1 X) |
| Color | Green |
| Latex Free | Yes |
| Material | Cotton, Fleece, NBR |
| Product Type | Chemical-resistant Gloves |
| Size | 8 |