missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rubisco activase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120014
This item is not returnable.
View return policy
Description
Positive Control: Mulberry leaf, rice, A. thaliana, tabacum. Subcellular Location: Plastid.
Ribulose bisphosphate carboxylase/oxygenase activase is an enzyme localized to chloroplasts which activates Rubisco by promoting ATP-dependent conformational changes. Alternative name: RuBisCo activase RCA. There are two forms of activase (alpha and beta) in some species (for example Arabidopsis, camelina, spinach, rice) and only one form in other species (tobacco, maize,Chlamydomonas). Alpha is about 46-47 kDa, beta is about 42 kDa. Species that have only one form have the beta form.
Specifications
| Rubisco activase | |
| Polyclonal | |
| Unconjugated | |
| RCA | |
| RCA | |
| Synthetic peptide within Arabidopsis thaliana Rubisco activase aa 190-233. | |
| 100 μL | |
| Primary | |
| Plant | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| 0 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 818558 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction