missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rubisco Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB110-81553
This item is not returnable.
View return policy
Description
Rubisco Polyclonal specifically detects Rubisco in Plant samples. It is validated for Western Blot.
Specifications
| Rubisco | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Rubisco | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000 | |
| 5-bisphosphate carboxylase/oxygenase small subunit N-methyltransferase I [Spinacia oleracea], EC 2.1.1.43, rbcL, Ribulose-1 | |
| 1-491 | |
| 0.05 mg | |
| Primary | |
| Plant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction