Learn More
RUFY1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13498-1-AP-20UL
 View more versions of this product
Product Code. 16881734
  • 145.00€ / 20µL
Description
This gene encodes a protein that contains a RUN domain and a FYVE-type zinc finger domain. The encoded protein binds to phosphatidylinositol-3-phosphate (PI3P) and plays a role in early endosomal trafficking, tethering and fusion through interactions with small GTPases including Rab4, Rab5 and Rab14. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
Specifications
RUFY1
Polyclonal
Unconjugated
RUFY1
FYVE finger protein EIP1, La binding protein 1, Rab4 interacting protein, RABIP4, RUFY1, ZFYVE12
Rabbit
Antigen Affinity Chromatography
RUO
216724, 360521, 80230
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BIJ7, Q96T51
Rufy1
RUFY1 Fusion Protein Ag4324