missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RUNX1 Recombinant Rabbit Monoclonal Antibody (3Y7V7)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558024
This item is not returnable.
View return policy
Description
Immunogen sequence: GTLVTVMAGN DENYSAELRN ATAAMKNQVA RFNDLRFVGR SGRGKSFTLT ITVFTNPPQV ATYHRAIKIT VDGPREPRRH RQKLDDQTKP GSLSFSERLS E.
RUNX1 belongs to the mammalian RUNX family of DNA binding proteins that regulate the expression of genes involved in cellular differentiation and cell cycle progression. RUNX1 is essential for hematopoietic stem cell generation in the vascular regions of the aorta, vitelline and umbilical arteries, yolk sac and placenta. RUNX1 is expressed in all tissues except brain and heart, with highest levels in thymus, bone marrow and peripheral blood.
Specifications
| RUNX1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q01196, Q03347 | |
| RUNX1 | |
| A synthetic peptide corresponding to a sequence within amino acids 101-200 of human RUNX1 (NP_001001890.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 3Y7V7 | |
| Unconjugated | |
| RUNX1 | |
| acute myeloid leukemia 1 protein; Acute myeloid leukemia 2 protein; acute myeloid leukemia 3 protein; acute myeloid leukemia gene 2; AML1; AML-1; aml1 oncogene; AML1-EVI-1; AML1-EVI-1 fusion protein; AML2; AML3; AMLCR1; CBF2alpha; CBFA1; CBFA2; CBFA3; CBF-alpha 2; CBF-alpha-1; CBF-alpha-2; CBF-alpha-3; CCD; CCD1; CLCD; core binding factor alpha 2; core-binding factor runt domain alpha subunit 2 (acute myeloid leukemia 1 oncogene); Core-binding factor subunit alpha-1; Core-binding factor subunit alpha-2; core-binding factor subunit alpha-3; core-binding factor, runt domain, alpha subunit 1; core-binding factor, runt domain, alpha subunit 2; core-binding factor, runt domain, alpha subunit 3; EVI-1; Oncogene AML-1; oncogene AML-2; Oncogene AML-3; OSF2; OSF-2; osteoblast-specific transcription factor 2; OTTHUMP00000108697; OTTHUMP00000108700; OTTHUMP00000108702; PEA2 alpha C; PEA2aA; PEA2-alpha A; PEA2-alpha B; PEA2-alpha C; PEBP2 alpha C; PEBP2A; Pebp2a2; PEBP2A3; PEBP2aA; PEBP2aB; PEBP2aC; PEBP2alpha; PEBP2-alpha A; PEBP2-alpha B; PEBP2-alpha C; Pebpa2b; polyomavirus enhancer-binding protein 2 alpha A subunit; Polyomavirus enhancer-binding protein 2 alpha B subunit; polyomavirus enhancer-binding protein 2 alpha C subunit; RUN1; runt domain, alpha subunit 2; runt related transcription factor 1; runt related transcription factor 2; runt related transcription factor 3; runt-related transcription factor 1; Runt-related transcription factor 2; runt-related transcription factor 3; Runx1; RUNX1/ZNF687 fusion; RUNX2; RUNX3; SL3/AKV core-binding factor alpha A subunit; SL3/AKV core-binding factor alpha B subunit; SL3/AKV core-binding factor alpha C subunit; SL3-3 enhancer factor 1 alpha A subunit; SL3-3 enhancer factor 1 alpha B subunit; SL3-3 enhancer factor 1 alpha C subunit; transcription factor; transcription factor AML2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 12394, 861 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction