missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX1 and RUNX1T1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-59160
This item is not returnable.
View return policy
Description
RUNX1 and RUNX1T1 Polyclonal specifically detects RUNX1 and RUNX1T1 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Chromatin Immunoprecipitation Sequencing.
Specifications
| RUNX1 and RUNX1T1 | |
| Polyclonal | |
| Western Blot 1:1000, ELISA 1:500, Chromatin Immunoprecipitation (ChIP) 4 uL/IP, Chromatin Immunoprecipitation Sequencing | |
| Acute myeloid leukemia 1 protein, AML1, AML1acute myeloid leukemia 1, AML1-EVI-1, AML1T1protein CBFA2T1, CBFA2, CBFA2acute myeloid leukemia 1 protein (oncogene AML-1), core-binding factor, alphasubunit10AMLCR1, CBFA2T1Cyclin-D-related protein, CDRMGC2796, Core-binding factor subunit alpha-2, core-binding factor, runt domain, alpha subunit 2; translocated to, 1; cyclinD-related, DKFZp564B213, Eight twenty one protein, ETOFLJ33145, EVI-1, MTG8acute myelogenous leukemia 1 translocation 1, cyclin-D related, PEBP2A2, Protein ETO, Protein MTG8, runt domain, alpha subunit 2, runt-related transcription factor 1, runt-related transcription factor 1; translocated to, 1 (cyclin D-related), Zinc finger MYND domain-containing protein 2, ZMYND2myeloid translocation gene on 8q22 | |
| The exact immunogen is propietary information. | |
| 100 μL | |
| Innate Immunity | |
| Human | |
| Antisera |
| Western Blot, ELISA, ChIP Assay | |
| Unconjugated | |
| Serum with 0.05% Sodium Azide | |
| Rabbit | |
| Whole Antiserum | |
| RUO | |
| Primary | |
| Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction