missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RUNX1/RUNX2/RUNX3 Recombinant Rabbit Monoclonal Antibody (23GB4800), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA553809

 View more versions of this product

Product Code. 30088055

  • 207.00€ / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Core binding factor (CBF) is a heterodimeric transcription factor that binds to the core element of many enhancers and promoters. The protein encoded by this gene represents the alpha subunit of CBF and is thought to be involved in the development of normal hematopoiesis. Chromosomal translocations involving this gene are welldocumented and have been associated with several types of leukemia. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RUNX1/RUNX2/RUNX3
Recombinant Monoclonal
375 μg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q01196, Q03347, Q08775, Q13761, Q13950, Q63046, Q64131, Q9Z2J9
RUNX1, Runx2, RUNX3
A synthesized peptide derived from human RUNX1/RUNX2/RUNX3 (400-453AA).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot
23GB4800
Unconjugated
Runx2
acute myeloid leukemia 1 protein; Acute myeloid leukemia 2 protein; acute myeloid leukemia 3 protein; acute myeloid leukemia gene 2; AKV core binding factor; AML1; AML1-EVI-1; AML1-EVI-1 fusion protein; AML2; Aml3; AMLCR1; Cbf; CBF2alpha; Cbfa1; Cbfa-1; CBFA2; Cbfa3; CBF-alpha-1; CBF-alpha-2; CBF-alpha-3; CCD; CCD1; CLCD; core binding factor alpha 1; core binding factor alpha 2; core binding factor alpha 3; core-binding factor runt domain alpha subunit 2 (acute myeloid leukemia 1 oncogene); core-binding factor subunit alpha-1; Core-binding factor subunit alpha-2; core-binding factor subunit alpha-3; core-binding factor, runt domain, alpha subunit 1; core-binding factor, runt domain, alpha subunit 2; core-binding factor, runt domain, alpha subunit 3; EVI-1; LS3; oncogene AML-1; Oncogene AML-2; oncogene AML-3; OSF2; OSF-2; Osteoblast-specific transcription factor 2; PEA2 alpha C; PEA2aA; PEA2-alpha A; PEA2-alpha B; PEA2-alpha C; PEBP2 alpha A; PEBP2 alpha C; Pebp2a; Pebp2a1; Pebp2a2; Pebp2a3; PEBP2aA; PEBP2aB; PEBP2aC; PEBP2alpha; PEBP2-alpha A; PEBP2-alpha B; PEBP2-alpha C; Pebpa2a; Pebpa2b; Polyomavirus enhancer-binding protein 2 alpha A subunit; polyomavirus enhancer-binding protein 2 alpha B subunit; Polyomavirus enhancer-binding protein 2 alpha C subunit; runt domain, alpha subunit 1; runt domain, alpha subunit 2; runt domain, alpha subunit 3; runt related transcription factor 1; runt related transcription factor 2; runt related transcription factor 3; runt-related transcription factor 1; Runt-related transcription factor 2; runt-related transcription factor 3; RUNX 2; Runx1; RUNX1/ZNF687 fusion; Runx2; RUNX-2; RUNX3; SL3/AKV core-binding factor alpha A subunit; SL3/AKV core-binding factor alpha B subunit; SL3/AKV core-binding factor alpha C subunit; SL3-3 enhancer factor 1 alpha A subunit; SL3-3 enhancer factor 1 alpha B subunit; SL3-3 enhancer factor 1 alpha C subunit; transcription factor; transcription factor AML2; transcription factor AML2/CBFA3
Rabbit
Affinity chromatography
RUO
12393, 12394, 12399, 156726, 367218, 50662, 860, 861, 864
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.