Learn More
RUNX2 Monoclonal Antibody (OTI3E12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813668
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
RUNX2 Monoclonal antibody specifically detects RUNX2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| RUNX2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q08775, Q13950, Q9Z2J9 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 12393, 367218, 860 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI3E12 | |
| Unconjugated | |
| RUNX2 | |
| Acute myeloid leukemia 3 protein; AKV core binding factor; Aml3; AML-3; Cbf; Cbfa1; Cbfa-1; CBF-alpha-1; CCD; CCD1; CLCD; core binding factor alpha 1; core binding factor alpha 1 subunit; core-binding factor subunit alpha-1; core-binding factor, runt domain, alpha subunit 1; LOW QUALITY PROTEIN: runt-related transcription factor 2; LS3; Oncogene AML-3; Osf2; OSF-2; osteoblast-specific transcription factor 2; PEA2aA; PEA2-alpha A; PEBP2 alpha A; PEBP2A; Pebp2a1; PEBP2aA; PEBP2-alpha A; Pebpa2a; Polyomavirus enhancer-binding protein 2 alpha A subunit; runt domain, alpha subunit 1; runt related transcription factor 2; runt-related transcription factor 2; RUNX 2; RUNX family transcription factor 2; Runx2; RUNX-2; SL3/AKV core-binding factor alpha A subunit; SL3-3 enhancer factor 1 alpha A subunit; transcription factor RUNX2; transcription factor RUNX2/CBFA1 | |
| Human recombinant protein fragment corresponding to amino acids 1-293 of human RUNX2 (NP_004339) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.