missing translation for 'onlineSavingsMsg'
Learn More

Ruxolitinib, Mikromol™

Product Code. 17982456
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17982456 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17982456 Supplier Mikromol Supplier No. MM3727.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ruxolitinib
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ruxolitinib
CAS 941678-49-5
Analyte or Component Names Ruxolitinib
Shipping Condition Room Temperature
Molecular Formula C17 H18 N6
Certifications/Compliance ISO 17034
Synonym (βR)-β-Cyclopentyl-4-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazole-1-propanenitrile, (3R)-3-Cyclopentyl-3-[4-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazol-1-yl]propanenitrile, (βR)-β-Cyclopentyl-4-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazole-1-propanenitrile, INC 424, INCB 018424, Ruxolitinib
InChI Formula InChI=1S/C17H18N6/c18-7-5-15(12-3-1-2-4-12)23-10-13(9-22-23)16-14-6-8-19-17(14)21-11-20-16/h6,8-12,15H,1-5H2,(H,19,20,21)/t15-/m1/s1
SMILES N#CC[C@H](C1CCCC1)n2cc(cn2)c3ncnc4[nH]ccc34
IUPAC Name (3R)-3-cyclopentyl-3-[4-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)pyrazol-1-yl]propanenitrile
Molecular Weight (g/mol) 306.36
Quantity 50 mg
Formula Weight 306.1593 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.